Learn More
Nelson Jameson Company 3M Quick Swabs, 250/case
Catalog No. NC1788568
Encompass
This ready-to-use environmental swab system consists of a 5-inch rayon-tipped swab which contains letheen neutralizing buffer to facilitate recovery of bacteria. The swabs can be used wet or dry to sample surfaces and quickly deliver 1.0 mL of sample onto a 3M Petrifilm™ Plate. Handy vinyl-covered wire rack which holds up to 50 swabs is sold separately.
Catalog No. NC1788568 Supplier Nelson Jameson Company Supplier No. 4603434 C
This ready-to-use environmental swab system consists of a 5-inch rayon-tipped swab which contains letheen neutralizing buffer to facilitate recovery of bacteria. The swabs can be used wet or dry to sample surfaces and quickly deliver 1.0 mL of sample onto a 3M Petrifilm™ Plate. Handy vinyl-covered wire rack which holds up to 50 swabs is sold separately.