Learn More
3M™ QX™ Safety Glasses
Catalog No. 19153632 Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Clear
Product Type:
QX 2000
Temple Style:
Adjustable
Catalog No. Lens Tint Product Type Temple Style
19-153-632 Clear QX 2000 Adjustable
1 options
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Catalog No. 19-153-632 Supplier 3M™ Supplier No. 7000002370
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Seamless lens for distortion-free vision
  • Sleek wraparound lens provides a wide field of vision and excellent side protection
  • Coated polycarbonate lens resists fogging, scratching, static and chemicals; provides 99.9% UV protection
  • Adjustable temples with changeable lengths
  • Lens is easily replaceable using the Easy Change™ system
  • Black frames
  • 3M Catalog# 12101-10000-20, 12115-10000-20, 12109-10000-20, 12100-10000-20
Sport Grip™ temples fit under headgear and ear muffs
Compliance
Meet ANSI™ Z87.1-2010 and CSA Z94.3 standards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Meet ANSI Z87.1-2010 and CSA Z94.3-2007 standards.
Lens Tint Clear
Product Type QX 2000
Frame Color Black
Temple Style Adjustable
Lens Material Polycarbonate