Learn More
3M 3M RAPID READOUT,1296 25/CS
Catalog No. NC1754435 Shop All 3M Products
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
3M Attest Rapid Readout Biological Indicator Test Pack, 1296, for 250F/121C Gravity and 270F/132C Vacuum Assisted Steam Sterilizers
Catalog No. NC1754435 Supplier 3M Supplier No. 7000002696/DEL
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item has a minimum qty of 2 per supplier requirements.
3M Attest Rapid Readout Biological Indicator Test Pack, 1296, for 250F/121C Gravity and 270F/132C Vacuum Assisted Steam Sterilizers