Learn More
3M™ RBE-CMP Breathing Tube Clamp
Catalog No. 19167750 Shop All 3M Products
Change view
Click to view available options
Model:
RBE-CMP
Catalog No. Model
19-167-750 RBE-CMP
1 options
Catalog No. 19-167-750 Supplier 3M™ Supplier No. 7010340259
Add to Cart
Add to Cart
Breathing tube clamp is used in conjunction with RBE-NM10 and RBE-L10 PAPR hood systems.
  • 3M Catalog# RBE-CMP
TRUSTED_SUSTAINABILITY
Specifications
Model RBE-CMP