Learn More
MSC 3M REMOTE ALARM
Catalog No. 5000684224
Encompass
The remote alarm and strobe provides an audible and visible indication outside the work area if the carbon monoxide (CO) level exceeds a preset level. It can be used with CO monitors in portable compressed air filter and regulator panels.
  • Audible remote alarm with 119 dBA output
  • Strobe light with 150,000 CP
  • Compatible with specific compressed air filter and regulator panels
  • Integrates with certain retrofit CO monitor kits
  • Supports 50 CFM and 100 CFM air flow capacity
  • Designed for supplied air systems
  • Suitable for MST portable and fixed panels
  • Not intrinsically safe
Catalog No. 50-006-84224 Supplier MSC Supplier No. 64258262
The remote alarm and strobe provides an audible and visible indication outside the work area if the carbon monoxide (CO) level exceeds a preset level. It can be used with CO monitors in portable compressed air filter and regulator panels.
  • Audible remote alarm with 119 dBA output
  • Strobe light with 150,000 CP
  • Compatible with specific compressed air filter and regulator panels
  • Integrates with certain retrofit CO monitor kits
  • Supports 50 CFM and 100 CFM air flow capacity
  • Designed for supplied air systems
  • Suitable for MST portable and fixed panels
  • Not intrinsically safe