Learn More
3M™ Replacement Fittings for Compressed Air Components: 0.37 Inch
Catalog No. 19813797 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Air high pressure hose, compressed air filter and regulator panel w-2806
Air low pressure hose w-3020
Product Type:
Hose Socket
Plug
Material:
Steel
Steel, Brass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Product Type Material
19-813-797 Air high pressure hose, compressed air filter and regulator panel w-2806 Hose Socket Steel, Brass
19-021-567 Air low pressure hose w-3020 Plug Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-813-797 Supplier 3M™ Supplier No. 7000130788
Add to Cart
Add to Cart
Designed for use with 3M compressed air components.
Replacement parts for compressed air components
  • High pressure, 3/8 inch MPT twist lock fitting for 3M™ Compressed Air Filter and Regulator Panel W-2806
  • Low pressure, 3/8 inch FPT twist lock steel and brass fitting for 3M™ Supplied Air Hose W-3020
  • 3M Catalog# W-3182-2 , 3M Catalog# W-3198-2
TRUSTED_SUSTAINABILITY
Specifications
Material Steel, Brass
Product Type Hose Socket
For Use With (Equipment) Air high pressure hose, compressed air filter and regulator panel w-2806
Includes Twist lock steel and brass fitting
Model W-3182-2
Length (English) 0.37 in.
Length (Metric) 0.93 cm