Learn More
3M™ Breathe-Easy™ Respirator Accessory, Airflow Indicator
Catalog No. 19823968 Shop All 3M Products
Change view
Click to view available options
Airflow Indicator for Breathe Easy PAPR Systems
Catalog No.
19-823-968 Airflow Indicator for Breathe Easy PAPR Systems
1 options
This item is not returnable. View return policy
Catalog No. 19-823-968 Supplier 3M™ Supplier No. 7000127066
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
For use only with the 3M™ Breathe Easy™ (BE) NIOSH CBRN approved tight fitting facepiece PAPR Systems. Airflow indicator is mounted at the hood end of the breathing tube.
  • 3M Catalog# AFI-100
TRUSTED_SUSTAINABILITY
Specifications
Disposable Reusable
Product Type Airflow Indicator
For Use With (Equipment) Breathe Easy™ PAPR Systems
Model AFI-100