Learn More
3M™ Breathe-Easy™ Respirator Accessory, Faceseal
Catalog No. 19019163 Shop All 3M Products
Change view
Click to view available options
Tychem QC Faceseal
Catalog No.
19-019163 Tychem QC Faceseal
1 options
Catalog No. 19-019163 Supplier 3M™ Supplier No. 7100010932
For use with Breathe Easy™, High Efficiency and Supplied Air Systems.
This product contains natural rubber. Natural rubber contains proteins that may cause reactions in persons who are allergic to latex or natural rubber products. 3M Catalog# 527-01-15R10
TRUSTED_SUSTAINABILITY
Specifications
Disposable Reusable
Material Tychem QC
Product Type Faceseal
For Use With (Application) Military Maintenance, Repair and Operation
For Use With (Equipment) Breathe Easy™ PAPR Systems
Model 527-01-15R10