Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Breathe-Easy™ Respirator Accessory,Turbo Powered Air Purifying Respirator Unit

Catalog No. 17987256 Shop All 3M Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-987-256 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-987-256 Supplier 3M™ Supplier No. 7000126342
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Replacement turbo blower unit for 3M™ Breathe Easy™ Systems.

  • Replacement part for 3M™ Breathe Easy™ Systems
  • Includes the turbo blower unit only
  • Full system kits include hood, motor blower, breathing tube, battery, airflow indicator, belt, and cartridge; some kits may have additional components
  • Headgear used as part of certain PAPRs can accommodate workers with limited facial hair and prescription eyewear
TRUSTED_SUSTAINABILITY

Specifications

Color Black
Product Type Motor Blower
For Use With (Application) Military maintenance, repair and operation
For Use With (Equipment) Breathe-Easy™ Respirators
Model 022-00-03R01
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.