Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Breathe-Easy™ Respirator Accessory, Airflow Indicator

Catalog No. 19120288 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Airstream™, Breath Easy™, and Powerflow™ PAPR Systems
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-120-288 Airstream™, Breath Easy™, and Powerflow™ PAPR Systems
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-120-288 Supplier 3M™ Supplier No. 7000131712
Add to Cart
Add to Cart

Used to indicate the airflow rate on 3M™ Breathe Easy™ and Powerflow™ Systems.

  • 3M Catalog# 520-01-21

TRUSTED_SUSTAINABILITY

Specifications

Color Transparent
Disposable Reusable
Product Type Airflow Indicator
Facepiece Type Full Facepiece
For Use With (Application) Health and Safety Testing, Indicate Airflow Rate
For Use With (Equipment) Airstream™, Breath Easy™, and Powerflow™ PAPR Systems
Model 520-01-21
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.