Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ PAPR Motor Blower Unit GVP-100

Catalog No. 19813769 Shop All 3M Products
Change view
Click to view available options
Motor blower; Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-813-769 Motor blower; Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-813-769 Supplier 3M™ Supplier No. 7000126091
This item is not returnable. View return policy

GVP-100 is a belt-mounted respirator unit that helps provide a level of air flow that surpasses NIOSH requirements.

  • GVP-100 is a belt-mounted PAPR motor blower unit
  • Depending on the headgear and cartridge, the airflow ranges from 7.0-9.5 CFM
  • Unit is lightweight and weighs only 1.1 lbs (0.5 kg), and has a life of up to 1000 hours
  • System is compatible with many different styles of headgear including lightweight hoods, bumpcaps, hard hats, helmets, and full facepiece respirators
  • Suggested applications include assembly, blasting, cleaning, metal repair and painting
TRUSTED_SUSTAINABILITY

Specifications

Color Black
Product Type Motor Blower
Flow Rate 7.0 to 9.5 CFM
For Use With (Application) Assembly, blasting, cleaning, metal repair and painting
For Use With (Equipment) Powered air purifying respirator system, lightweight hoods, bumpcaps, hard hats, helmets and full facepiece respirator
Includes Loose fitting headgear, battery powered blower
Model GVP-100
Weight (English) 1.1 lb.
Weight (Metric) 0.49 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.