Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Replacement Parts for Air-Mate™ Belt Mounted High Efficiency PAPR

Catalog No. 18038 Shop All 3M Products
Change view
Click to view available options
Nylon belt
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-038 Nylon belt
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-038 Supplier 3M™ Supplier No. 7000029681

Replacement parts for Air-Mate high efficiency powered air purifying respirator systems

  • Nylon belt
  • Head cover is white; Large size
  • 3M Catalog# 021-41-02R01 , 3M Catalog# 451-02-01R01

18033: Air-Mate PAPR Unit
18035: Airflow Indicator; Black; Nylon
18043: HE filter; White; NIOSH-approved
19-039-795: Head Cover; Large; White

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Disposable Reusable
Material Nylon
Product Type Belt
For Use With (Application) Assembly, Machine Operation
For Use With (Equipment) AirMate™ High Efficiency PAPR System
Model 021-41-02R01
Length (English) 59 in.
Length (Metric) 149.86 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.