Learn More
3M™ Replacement Parts for Air-Mate™ Belt Mounted High Efficiency PAPR
Catalog No. 18035 Shop All 3M Products
Change view
Click to view available options
Airflow indicator
Catalog No.
18-035 Airflow indicator
1 options
Catalog No. 18-035 Supplier 3M™ Supplier No. 0211400R01
Replacement parts for Air-Mate high efficiency powered air purifying respirator systems
  • Nylon belt
  • Head cover is white; Large size
  • 3M Catalog# 021-41-02R01 , 3M Catalog# 451-02-01R01
18033: Air-Mate PAPR Unit
18035: Airflow Indicator; Black; Nylon
18043: HE filter; White; NIOSH-approved
19-039-795: Head Cover; Large; White
TRUSTED_SUSTAINABILITY
Specifications
Disposable Reusable
For Use With (Application) Ensure That The Breathing Tube is Connected to The PAPR Unit and That The HE Filter and Battery Pk. Are in Place
For Use With (Equipment) AirMate™ High Efficiency PAPR System
Model 021-14-00R01