Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Replacement Parts for Air-Mate™ Belt Mounted High Efficiency PAPR

Catalog No. 18043 Shop All 3M Products
Change view
Click to view available options
High efficiency filter with gasket
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-043 High efficiency filter with gasket
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-043 Supplier 3M™ Supplier No. 7000005590

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Replacement parts for Air-Mate high efficiency powered air purifying respirator systems

  • Nylon belt
  • Head cover is white; Large size
  • 3M Catalog# 021-41-02R01 , 3M Catalog# 451-02-01R01

18033: Air-Mate PAPR Unit
18035: Airflow Indicator; Black; Nylon
18043: HE filter; White; NIOSH-approved
19-039-795: Head Cover; Large; White

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance NIOSH Approved
Color Magenta
Disposable Reusable
Product Type HEPA Filter
For Use With (Application) Protection Against Dusts, Musts, Fumes, Asbestos, Radionuclides and Radon Daughter
For Use With (Equipment) AirMate™ High Efficiency PAPR System
Includes Gasket
Model 451-02-01R01
Weight (English) 0.54 lb.
Weight (Metric) 0.24 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.