Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Tinted Visor for Airstream Headgear-Mounted PAPR

Catalog No. 19018990 Shop All 3M Products
Change view
Click to view available options
Visor; Gray tinted
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-018990 Visor; Gray tinted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-018990 Supplier 3M™ Supplier No. 7000126351

missing translation for 'validMainframeMsg'

Replacement part for use with 3M™ Breathe Easy, Airstream High Efficiency and Supplied Air Systems.

Gray tinted visor for use with Airstream Headgear-Mounted PAPR.

  • 3M Catalog# 060-75-02R10

TRUSTED_SUSTAINABILITY

Specifications

Color Gray
Disposable Reusable
Product Type Visor
For Use With (Application) Assembly, Machine Operation
For Use With (Equipment) Breathe Easy™, Airstream™ PAPR Systems
Model 060-75-02R10
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.