Learn More
3M™ Versaflo™ TR-300 / Speedglas™ TR-300 SG PAPR Accessory, Airflow Indicator
Catalog No. 19039577 Shop All 3M Products
Change view
Click to view available options
Airflow indicator
Catalog No.
19-039-577 Airflow indicator
1 options
Catalog No. 19-039-577 Supplier 3M™ Supplier No. 7100009677
The TR-971 air flow indicator may be used to confirm air flow levels of the TR-600 series PAPR. The TR-971 is designed to enable testing at various elevations and temperatures within the operational range of the PAPR system.
The TR-971 air flow indicator may be used to confirm air flow levels of the TR-600 series PAPR. The TR-971 is designed to enable testing at various elevations and temperatures within the operational range of the PAPR system. See the User Instructions for details regarding how to use the TR-971.
  • Vortemp Air-Heating valve
  • ADF not included
  • 3M Catalog# TR-970
TRUSTED_SUSTAINABILITY
Specifications
Disposable Reusable
Product Type Airflow Indicator
For Use With (Application) Ideal to Confirm Air Flow Levels and to Enable Testing at Various Elevations and Temperatures Within The Operational Range of The Powered Air Purifying Respirator System
For Use With (Equipment) Versaflo TR-300, Speedglas TR-300 SG PAPR System
Model TR-970