Learn More
Enterprise Technology Solutions 3M Respirator,Particu.,P95,M-Noseclip,CarbonLayer,1Size,8/CT,WE
SDP
Catalog No. 5000827955
Encompass_Preferred
Particulate Respirator 8577 provides P95-level protection against oil and non-oil based aerosols. Added carbon-layering in the filter helps minimize organic vapor bleed-through. Lightweight design and adjustable M-nose clip enhance comfort levels, particularly over extended use. Respirators are ideal for hot and humid work environments. One size fits all.
Catalog No. 50-008-27955 Supplier Enterprise Technology Solutions Supplier No. MMM8577CT
Particulate Respirator 8577 provides P95-level protection against oil and non-oil based aerosols. Added carbon-layering in the filter helps minimize organic vapor bleed-through. Lightweight design and adjustable M-nose clip enhance comfort levels, particularly over extended use. Respirators are ideal for hot and humid work environments. One size fits all.