Learn More
MSC 3M RESPIRATOR RETAINER
Catalog No. 5000686171
Encompass
The 3M™ Filter Retainer 501 is a component designed for respiratory protection systems. It holds particulate filters in place on various respirators and cartridges, including 3M™ Respirators 5000 Series, 3M™ Cartridges 6000 Series, and 3M™ Filter Adapter 603, as well as 3M™ 7000 Series Gas and Vapour cartridges.
  • Approved system component
  • Designed to hold particulate filters
  • Compatible with multiple respirator and cartridge series
  • Constructed from polypropylene
  • Translucent white color
  • Approved to AS/NZS1716 standard
Catalog No. 50-006-86171 Supplier MSC Supplier No. 00324475
The 3M™ Filter Retainer 501 is a component designed for respiratory protection systems. It holds particulate filters in place on various respirators and cartridges, including 3M™ Respirators 5000 Series, 3M™ Cartridges 6000 Series, and 3M™ Filter Adapter 603, as well as 3M™ 7000 Series Gas and Vapour cartridges.
  • Approved system component
  • Designed to hold particulate filters
  • Compatible with multiple respirator and cartridge series
  • Constructed from polypropylene
  • Translucent white color
  • Approved to AS/NZS1716 standard