Learn More
Enterprise Technology Solutions 3M Respirators,N95,2Strap,NonOil,StandardSize,20/BX,8/CT,WE
SDP
Catalog No. 5000827909
Encompass_Preferred
8000 Cup Series Particulate Respirators are lightweight, disposable respirators designed to provide convenient, reliable respiratory protection. Adjustable nose clip and two-strap design ensure a more secure seal against your face. Advanced electrostatic media is designed for ease of breathing. Masks are suggested for grinding, sanding, sweeping and other non-oil based operations.
Catalog No. 50-008-27909 Supplier Enterprise Technology Solutions Supplier No. MMM8210CT
8000 Cup Series Particulate Respirators are lightweight, disposable respirators designed to provide convenient, reliable respiratory protection. Adjustable nose clip and two-strap design ensure a more secure seal against your face. Advanced electrostatic media is designed for ease of breathing. Masks are suggested for grinding, sanding, sweeping and other non-oil based operations.