Learn More
MSC 3M RESPIRATORY PROTECTION
Catalog No. 5000802973
Encompass
This sensitivity solution is designed for qualitative respiratory fit testing, providing a distinct bitter taste for accurate assessment.
  • Solution type: sensitivity solution
  • Container type: bottle
  • Container size: 55.0 ml
  • Country of origin: United States
Catalog No. 50-008-02973 Supplier MSC Supplier No. 06418610
This sensitivity solution is designed for qualitative respiratory fit testing, providing a distinct bitter taste for accurate assessment.
  • Solution type: sensitivity solution
  • Container type: bottle
  • Container size: 55.0 ml
  • Country of origin: United States