Learn More
MSC 3M RESPIRATORY PROTECTION
Catalog No. 5000803470
Encompass
This sensitivity solution is designed for qualitative fit testing of respirators, aiding organizations in meeting OSHA requirements by checking the face-to-respirator seal. It is a sweet-type solution formulated for industrial and occupational use.
  • Sweet taste for sensitivity testing
  • Used with Qualitative Fit Test Apparatus
  • Aids in meeting OSHA fit testing requirements
  • Colourless, odourless liquid
  • Aqueous solution containing less than 1% sodium saccharin
  • Packaged in a 55 mL bottle
Catalog No. 50-008-03470 Supplier MSC Supplier No. 06418586
This sensitivity solution is designed for qualitative fit testing of respirators, aiding organizations in meeting OSHA requirements by checking the face-to-respirator seal. It is a sweet-type solution formulated for industrial and occupational use.
  • Sweet taste for sensitivity testing
  • Used with Qualitative Fit Test Apparatus
  • Aids in meeting OSHA fit testing requirements
  • Colourless, odourless liquid
  • Aqueous solution containing less than 1% sodium saccharin
  • Packaged in a 55 mL bottle