Learn More
MSC 3M RESPIRATORY PROTECTION
Catalog No. 5000769432
Encompass
This low-pressure SAR (Supplied Air Respirator) supply hose is 50 ft. long with a 1/2 in. inside diameter. It is designed for use with compatible low pressure compressed air systems, providing reliable air supply in respiratory protection applications.
  • Straight hose type
  • Low pressure type
  • Overall length of 50.0 ft.
  • Inner diameter of 1/2 in.
  • Compatible with low pressure compressed air systems
Catalog No. 50-007-69432 Supplier MSC Supplier No. 06418792
This low-pressure SAR (Supplied Air Respirator) supply hose is 50 ft. long with a 1/2 in. inside diameter. It is designed for use with compatible low pressure compressed air systems, providing reliable air supply in respiratory protection applications.
  • Straight hose type
  • Low pressure type
  • Overall length of 50.0 ft.
  • Inner diameter of 1/2 in.
  • Compatible with low pressure compressed air systems