Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ PAPR RBE System Accessory, Belt

Catalog No. 19167747 Shop All 3M Products
Change view
Click to view available options
RBE belt
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-167-747 RBE belt
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-167-747 Supplier 3M™ Supplier No. 7000126825
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with RBE-NM10, RBE-L10, RBE-NMB and RBE-LB PAPR systems.

  • Compatible with RBE-NM10, RBE-L10, RBE-NMB and RBE-LB Systems
  • Fits waist size up to 60 inches
  • Black
  • 3M Catalog# RBE-BLT

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Disposable Reusable
Product Type Belt
For Use With (Equipment) RBE-NM10, RBE-L10, RBE-NMB, and RBE-1 B Powered Air Purifying Respirator System
Model RBE-BLT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.