Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Breathe Easy™ PAPR Hood System Accessory, Breathing Tube

Catalog No. 19167748 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
RBE-NM10 and RBE-L10 PAPR Hood Systems
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-167-748 RBE-NM10 and RBE-L10 PAPR Hood Systems
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-167-748 Supplier 3M™ Supplier No. 7010384723
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Breathing tube for use with RBE-NM10 and RBE-L10 PAPR systems.

  • Compatible with RBE-NM10 and RBE-L10 systems
  • Back-mounted
  • Black
  • 3M Catalog# RBE-BTH

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Product Type Breathing Tube
For Use With (Equipment) RBE-NM10 and RBE-L10 PAPR Hood Systems
Model RBE-BTH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.