Learn More
MSC 3M RPL LENS ASSEMBLY
Catalog No. 5000686668
Encompass
This lens assembly is an approved replacement part recommended for 3M Full Facepiece 6000 Series respirators. This assembly is designed to extend the service life of the facepiece, offering potential cost savings while maintaining original service performance.
  • Approved replacement part
  • Recommended for 6000 Series full facepiece respirators
  • Extends service life of facepiece
  • Can result in respiratory equipment cost savings
  • Maintains original service performance of facepiece
  • Constructed from polycarbonate material
Catalog No. 50-006-86668 Supplier MSC Supplier No. 66686544
This lens assembly is an approved replacement part recommended for 3M Full Facepiece 6000 Series respirators. This assembly is designed to extend the service life of the facepiece, offering potential cost savings while maintaining original service performance.
  • Approved replacement part
  • Recommended for 6000 Series full facepiece respirators
  • Extends service life of facepiece
  • Can result in respiratory equipment cost savings
  • Maintains original service performance of facepiece
  • Constructed from polycarbonate material