Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ Chin Strap for S-950 Premium Head Suspension

Catalog No. 19805544 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Premium Head Suspension S-950
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-805-544 Premium Head Suspension S-950
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-805-544 Supplier 3M™ Supplier No. 7000127464
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with S-950 Premium Head Suspension.

  • For use with S-950 Premium Head Suspension
  • 3M Catalog# S-958

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OSHA
Color Gray
Disposable Reusable
Product Type Chin Strap
For Use With (Equipment) Premium Head Suspension S-950
Model S-958
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.