Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ Premium Head Suspension for S-series Hood Assembly

Catalog No. 19805604 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
S-Series Hood Assembly
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-805-604 S-Series Hood Assembly
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-805-604 Supplier 3M™ Supplier No. 7100005445
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Resusable head suspension is for use with the S-Series Hood Assembly.

  • Reusable
  • Color: Gray
  • 3M Catalog# S-950

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OSHA
Color Gray
Disposable Reusable
Product Type Head Suspension
For Use With (Application) Assembly, Cleaning, Electrical, Facility Maintenance
For Use With (Equipment) S-Series Hood Assembly
Model S-950
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.