Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ Peel-Off Visor Cover for Large S-Series Hood and Headcovers

Catalog No. 19805622 Shop All 3M Products
Change view
Click to view available options
Peel-off visor cover; Medium/Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-805-622 Peel-off visor cover; Medium/Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-805-622 Supplier 3M™ Supplier No. 7000127434
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Peel-off visor cover protects hoods and head covers from contaminants and scratches. For use with large assemblies only.

  • Transparent covering to hood
  • Protect from contaminants and scratches
  • 3M Catalog# S-920L

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Disposable Reusable
Product Type Lens Cover
Size Large
For Use With (Application) Protects Visor From Contaminants and Scratches
For Use With (Equipment) Large Headcovers and Hoods, S133L, S133LB, S333L, S433L, S533L
Model S-920 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.