Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ Peel-Off Visor Cover for S-Series Hood Assembly

Catalog No. 19805624 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
S-Series Hoods and Headcovers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-805-624 S-Series Hoods and Headcovers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-805-624 Supplier 3M™ Supplier No. 7100138630
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Peel-off visor cover is for use with S-Series hoods and headcovers to protect from contaminants and scratches.

  • Transparent covering to hood
  • Protect from contaminants and scratches
  • 3M Catalog# S-922

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Disposable Reusable
Product Type Lens Cover
For Use With (Application) Protects Visor From Contaminants and Scratches
For Use With (Equipment) S-Series Hoods and Headcovers
Model S-922
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.