Learn More
3M™ Safety Splash Goggle
Catalog No. 19065423 Shop All 3M Products
Change view
Click to view available options
Lens Coating:
Anti-fog
Catalog No. Lens Coating
19-065-423 Anti-fog
1 options
This item is not returnable. View return policy
Catalog No. 19-065-423 Supplier 3M™ Supplier No. 7000002378
  • $15.75
    $8.65 / Each
    Save $7.10
    45% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $157.00
    $86.65 / Case of 10 EA
    Save $70.35
    45% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Economical goggle 334AF protective eyewear offers indirect anti-fog venting, an adjustable elastic strap, and polycarbonate construction with over-the-glass coverage. Indoor or outdoor use safety goggles.
These safety splash goggles are a dependable and economical choice that offer a sturdy polycarbonate lens, adjustable elastic strap and a wide opening to fit over most prescription eyewear. It is built with an indirect venting system that circulates air and helps minimize fogging, allowing for use in varied environments and conditions.
  • Economical goggle offers indirect venting and anti-fog coating for reduced fogging
  • Sturdy polycarbonate lens
  • Wide opening to fit over prescription eyewear
  • 3M Catalog# 40661-00000-10
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Lens Tint Clear
Product Type Splash Goggles
No. per Case 10/Cs.
Model 40661-00000-10
Material Polycarbonate