Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Safety-Walk™ 610 Slip-Resistant Floor Tread

Catalog No. 19813704 Shop All 3M Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-813-704 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-813-704 Supplier 3M™ Supplier No. 7100009879

missing translation for 'validMainframeMsg'

Mineral-coated, slip-resistant material features a highly durable surface

  • Low-profile design reduces trip hazard
  • Substrate material: Plastic film
  • Surface material: Mineral
  • Adhesive
  • Heavy duty
  • 3M Catalog# 610

Compliance

Eu Rohs Compliant

TRUSTED_SUSTAINABILITY

Specifications

Dimensions (L x W) 60 ft. x 24 in.
Temperature Range (English) 40°F to 175°F
For Use With (Application) Automotive, Ladder, Machinery, Stairs
Length (Metric) 18.2 m
Width (English) 24 in.
Temperature Range (Metric) 4.4°C to 79.4°C
Width (Metric) 60.9 cm
Packaging Quantity 1 Ea.
Length (English) 60 ft.
Type Safety-Walk 610 Slip-Resistant Tapes and Treads
Color Black
Adhesive Type Pressure Sensitive Adhesive
Material Mineral-coated Plastic
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.