Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Safety-Walk™ General Purpose Floor Tape

Catalog No. 19040891 Shop All 3M Products
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-040-891 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-040-891 Supplier 3M™ Supplier No. 7000126472

missing translation for 'validMainframeMsg'

Mineral-coated, slip-resistant material features a highly durable surface

  • Low-profile design helps reduce trip hazards
  • Form type: Roll
  • Substrate material: Plastic film
  • Surface material: Mineral
  • Tread grade: Heavy duty
  • Color: Safety Yellow
  • W x L: 4 in. x 60 ft.
  • 3M Catalog# 630-B
TRUSTED_SUSTAINABILITY

Specifications

Dimensions (L x W) 60 ft. x 4 in.
Color Yellow
Packaging Quantity 1 Ea.
For Use With (Application) Automotive, Ladder, Machinery, Stairs
Width (English) 4 in.
Width (Metric) 10.16 cm
Type Floor Tape
Length (English) 60 ft.
Length (Metric) 18.2 m
Material Mineral-coated Plastic
Certifications/Compliance EU RoHS
Adhesive Type Pressure Sensitive Adhesive
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.