Learn More
Enterprise Technology Solutions 3M SafetyHelmet,SecureFit,4Point,Adjustable,Uvicator,20/CT,BE
SDP
Catalog No. 5000828008
Encompass_Preferred
Help give your workers an excellent blend of comfort, style and safety with this SecureFit Hard Hat H-700 Series. Suspension system incorporates exclusive patented SecureFit Pressure Diffusion Technology and sits low on the head for greater comfort and security. Comfort pad on back of suspension is designed to distribute force and increase ease of wear. Customize your fit with multiple levels of height and head-shape adjustments. Premium washable and replaceable sweatband with moisture-wicking material is designed to help keep workers cool and dry. Uvicator sensor lets wearers know when to replace hard hat due to UV exposure. Hard hats meet ANSI Z89.1 Safety Compliance Standards.
Catalog No. 50-008-28008 Supplier Enterprise Technology Solutions Supplier No. MMMH703SFRUVCT
Help give your workers an excellent blend of comfort, style and safety with this SecureFit Hard Hat H-700 Series. Suspension system incorporates exclusive patented SecureFit Pressure Diffusion Technology and sits low on the head for greater comfort and security. Comfort pad on back of suspension is designed to distribute force and increase ease of wear. Customize your fit with multiple levels of height and head-shape adjustments. Premium washable and replaceable sweatband with moisture-wicking material is designed to help keep workers cool and dry. Uvicator sensor lets wearers know when to replace hard hat due to UV exposure. Hard hats meet ANSI Z89.1 Safety Compliance Standards.