Learn More
Electron Microscopy Sciences Scotch™ 810 Magic Tape 3/4" 72 YD
SDP
Catalog No. 5028510
Encompass_Preferred
The ever popular tape that is invisible when applied. Can be written on. Resists yellowing, cracking, and moisture. Ideal for mending, slicing, and many other permanent applications. 3" core. 3" core.
Catalog No. 50-285-10 Supplier Electron Microscopy Sciences Supplier No. 77301
The ever popular tape that is invisible when applied. Can be written on. Resists yellowing, cracking, and moisture. Ideal for mending, slicing, and many other permanent applications. 3" core. 3" core.