Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Scotch™ Brand H-122 Box-Sealing Tape Dispenser

Catalog No. 19047273 Shop All 3M Products
Change view
Click to view available options
Model:
H122
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
19-047-273 H122
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-047-273 Supplier 3M™ Supplier No. 7000005398
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Durable, high-impact plastic construction

  • Compact and lightweight for easy, accurate, handheld sealing
  • Built-in braking device allows tighter, more precise seals
  • Holds 60 yd. (55m) rolls up to 2 in. wide (5cm)
  • 3M Catalog# H122

TRUSTED_SUSTAINABILITY

Specifications

Product Type Box Sealing Tape Dispenser
Color Gray
Material Plastic
Capacity 1 Roll, 2 in. W x 60 yd. L
For Use With (Application) Lightweight, compact, hand-held dispenser with built-in brake for tighter, more precise seals
Model H122
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.