Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Scotch™ Brand H-180 Box Sealing Roll-On Dispenser

Catalog No. 19047274 Shop All 3M Products
Change view
Click to view available options
Model:
H180
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
19-047-274 H180
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-047-274 Supplier 3M™ Supplier No. 7000042997
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lightweight, handheld tape

  • Features a comfortable pistol grip for easy operation
  • Adjustable tape brake allows control of tension and delivery
  • Holds tape rolls up to 60 yd. long (55m) and up to 2 in. wide (5cm)
  • 3M Catalog# 19008

TRUSTED_SUSTAINABILITY

Specifications

Product Type Box Sealing Tape Dispenser
Color Tan, Gray
Material Metal, Plastic
Capacity 1 Roll, 2 in.W x 60 yd. L
For Use With (Application) Lightweight, easy-to-operate, allows operator to apply 2 inch wide tape for box sealing tape applications
Model H180
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.