Learn More
Electron Microscopy Sciences Scotch™ Lithographers Tape
SDP
Catalog No. 5028507
Encompass_Preferred
This ruby-red transparent tape is for use in opaquing, stripping, and edging on film materials. 3" core.
Catalog No. 50-285-07 Supplier Electron Microscopy Sciences Supplier No. 77500
This ruby-red transparent tape is for use in opaquing, stripping, and edging on film materials. 3" core.