Learn More
3M™ 3M™ SecureFit™ 500 Series Safety Glasses
Catalog No. 19072296 Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Clear
Gray
Gray I/O
Lens Coating:
Anti-fog, Scratch-resistant
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Lens Coating
19-072-296 Clear Anti-fog, Scratch-resistant
19-072-253 Gray Anti-fog, Scratch-resistant
19-072-274 Gray I/O Anti-fog, Scratch-resistant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-072-296 Supplier 3M™ Supplier No. 7100228736
Add to Cart
Add to Cart
This item is not returnable. View return policy
3M™ SecureFit™ Protective Eyewear 500 Series features a stylized lens shape with temples that automatically self-adjust to help provide a secure and comfortable fit.
  • Adjustable ratchet temples allow user to adjust lens for facial features and eyeglass position
  • Sporty design with contemporary lens shape
  • SecureFit™ Pressure Diffusion Temple technology design features slotted ribs that provide comfortable pressure equalization. The temples flex and self-adjust to the size of wearer's head.
  • Workers can feel confident wearing 3M eye protection that meets the requirements of ANSI Z87.1-2015 Standard as an Impact Rated Spectacle
  • Polycarbonate lenses absorb 99.9% of UVA and UVB rays.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Scratch-resistant
Certifications/Compliance Z87+ U6
Lens Tint Clear
Product Type Protective Eyewear
For Use With (Application) Assembly, Machining, Remediation, Primary Metals, Power Fastening, Pouring/Casting, Pharmaceuticals, Painting, Oil and Gas, Mining, Military, Military Tactical, Military Maintenance, Repair and Operation (MRO), Masonary, Law Enforcement, Machine Operations, Automotive, Grinding, General Manufacturing, Food and Beverage Manufacturing, Food & Beverage, Facility Maintenance, Drilling, Demolition, Construction, Chiseling, Chemicals, Chemical Handling, Aviation, Sub Applications
Frame Color Clear
No. per Case 20/Cs.
Temple Style 3M Pressure Diffusion Temple Technology, Adjustable Temples
Temple Color Black
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Frameless