Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Belt-Mounted PAPR Respirator System Accessory, Shower Cover

Catalog No. 19813774 Shop All 3M Products
Change view
Click to view available options
Shower cover
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-813-774 Shower cover
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-813-774 Supplier 3M™ Supplier No. 7000126223

missing translation for 'validMainframeMsg'

Shower cover is an optional component for 3M™ belt-mounted PAPR systems.

  • GVP series
  • Color: Clear
  •  
  • 3M Catalog# GVP-119

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Disposable Reusable
Product Type Shower Cover
For Use With (Equipment) Self Contained Breathing Apparatus
Model GVP-119
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.