Learn More
Electron Microscopy Sciences Scotch™ Silver Tape 3/8" 72 YD
SDP
Catalog No. 5028515
Encompass_Preferred
Good for making glass knife boats as well as for hanging, protecting and stripping. 3" core. 3/8" width.
Catalog No. 50-285-15 Supplier Electron Microscopy Sciences Supplier No. 77700
Good for making glass knife boats as well as for hanging, protecting and stripping. 3" core. 3/8" width.