Learn More
Quality Biological Inc Sodium Acetate (3 M), pH 5.2, 100 mL
SDP
Supplier:  Quality Biological Inc 351035721EA
Encompass_Preferred
Sodium Acetate, 3M pH 5.2 is a common molecular biology reagent. ¿ All production lots are tested to ensure that they are free of Dnase, Rnase & Proteases. ¿ It is often used in buffer systems to maintain the pH of a given solution. ¿ Filtered through a 0.22 ¿m membraneDnase, Rnase, and Protease tested ¿ Applications: Sodium Acetate Solution 3M, pH 5.2 is a 0.22 ¿m membrane filtered, sterile, nuclease-free solution of sodium acetate. ¿ This solution is routinely used in DNA and RNA precipitation, elution of proteins and other uses in molecular biology applications. ¿ Storage Conditions: 15¿C to 30¿C ¿ Shipping condition: ambient temperature ¿ Sterile Filtered ¿ DNase RNase and Protease tested ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ ¿ Max Shelf Life: 36 months
Catalog No. 50-751-7299