Learn More
3M™ Solus™ 1000 Series Safety Glasses
Catalog No. 19072243 Shop All 3M Products
Change view
Click to view available options
Style:
Classic
Foam lined gasket with strap
Lens Tint:
Clear
Gray
Gray I/O
Indoor/Outdoor Vermilion
Vermillion
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Style Lens Tint
19-072-243 Classic Gray
19-072-259 Foam lined gasket with strap Clear
19-072-302 Foam lined gasket with strap Indoor/Outdoor Vermilion
19-072-306 Foam lined gasket with strap Gray
19-072-307 Classic Gray I/O
19-072-311 Classic Clear
19-072-266 Foam lined gasket with strap Vermillion
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-072-243 Supplier 3M™ Supplier No. 7100221210
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
3M™ Solus™ 1000 Series Safety Glasses feature a sporty low profile and are available in multiple frame colors and lens tints with Scotchgard™ Anti-fog Protector Coating.
  • Contemporary styling with soft nosebridge for added comfort
  • Scotchgard™ Protector Anti-Fog Coating resists fogging longer than traditional anti-fog coatings, up to 25 washings with water (Based on 3M internal testing per EN168 test method when compared with traditional anti-fog coatings.)
  • Removable temples with optional elastic strap/headband
  • Optional removable foam-lined gasket has vents to reduce fogging and helps limit eye exposure to nuisance dust while providing added comfort and cushioning.
  • Polycarbonate lens absorbs 99.9% UVA and UVB rays
  • Meets the requirements of ANSI Z87.1-2015 Standard as an Impact Rated spectacle
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Scratch-resistant
Certifications/Compliance ANSI Z87.1-2015
Lens Tint Gray
Product Type Safety Glasses
For Use With (Application) Assembly, Machining, Transportation, Sub Applications, Sawing, Riveting, Remediation, Primary Metals, Power Fastening, Pouring/Casting, Pharmaceuticals, Oil and Gas, Mining, Masonary, Law Enforcement, Machine Operations, Automotive, Grinding, General Manufacturing, Food and Beverage Manufacturing, Food & Beverage, Facility Maintenance, Drilling, Demolition, Construction, Chiseling, Chemicals, Chemical Handling, Aviation, UV Protection
No. per Case 20/Cs.
Style Classic
Temple Color Black
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection UV400
Frame Type Frameless