Learn More
3M™ 3M™ Solus™ CCS Series
Catalog No. 03052852 Shop All 3M Products
Change view
Click to view available options
Lens Coating:
Anti-fog
Catalog No. Lens Coating
03-052-852 Anti-fog
1 options
Catalog No. 03-052-852 Supplier 3M™ Supplier No. 7100196393
  • $641.00
    $400.65 / Case of 20
    Save $240.35
    37% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Designed to keep eyewear and earplugs attached, untangled and ready to use, so your team has the convenient, comfortable protection they need to focus on the jobs at hand. All models feature 3M™ Scotchgard™ Protector Anti-fog coating. A foam gasket is optional with small vents in the gasket help minimize fogging.
  • Features a Cord Control System (CCS) that helps keep eyewear and earplugs attached, untangled and ready to use
  • 3M™ Scotchgard™ Protector Anti-Fog Coating resists fogging longer than traditional anti-fog coatings through up to 25 washings with water* and helps provide scratch resistance in tough work environments. *Based on 3M internal testing per EN168 test method when compared with traditional anti-fog coatings.
  • Contemporary styling with soft nosebridge for added comfort
  • Small vents in the optional foam gasket help minimize fogging.
  • U6 rated polycarbonate lenses absorb 99.9% of UVA, UVB, and UVC rays between 200 nm and 380 nm
  • Workers can feel confident wearing 3M eye protection that meets the Impact-Rated Protector requirement of the ANSI-ISEA Z87.1 Standard
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Shape Wraparound
Certifications/Compliance ANSI Z87.1-2015, CSA Z94.3, U6
Lens Tint Clear
Product Type Protective Eyewear
For Use With (Application) Assembly, Automotive, Aviation, Chemical Handling, Chemicals, Chiseling, Construction, Demolition, Drilling, Facility Maintenance, Facility Maintenance, Food & Beverage, Food and Beverage Manufacturing, General Manufacturing, Grinding, Law Enforcement, Machine Operations, Machining, Masonary, Metal Fabrication, Mining, Oil and Gas, Pharmaceuticals, Pouring/Casting, Power Fastening, Primary Metals, Remediation, Riveting, Sawing, Transportation, UV Protection
Frame Color Green
No. per Case 20/Cs.
Model SCCS01SGAF-GRN-F
Style Foam Gasket
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Nylon
UV Protection UV400
Frame Type Frameless