Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ P95 Particulate Respirator, 8271

Catalog No. 19002709 Shop All 3M Products
Change view
Click to view available options
Stapled strap attachment; Exhalation Valve; M-nose clip
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-002-709 Stapled strap attachment; Exhalation Valve; M-nose clip
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Do not exceed maximum use concentration established by regulatory standards. If respirator becomes damaged, soiled, or breathing becomes difficult, leave the contaminated area and dispose of the respirator. Before each use of the product, please inspect for any rips, damage, tears, etc. These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Catalog No. 19-002-709 Supplier 3M™ Supplier No. 7000002047
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Do not exceed maximum use concentration established by regulatory standards. If respirator becomes damaged, soiled, or breathing becomes difficult, leave the contaminated area and dispose of the respirator. Before each use of the product, please inspect for any rips, damage, tears, etc. These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.

Provides reliable respiratory protection of at least 95 percent filtration efficiency against certain oil and non oil based particles. Soft inner material provides added comfort while the cup shape design helps make the respirator feel spacious and durable.

  • NIOSH approved P95 rating
  • Adjustable M-noseclip
  • Braided headbands, staple attachment
  • Foam faceseal
  • Cool Flow™ Exhalation Valve
TRUSTED_SUSTAINABILITY

Specifications

Aerosol Type P - Oil-Proof
Assigned Protection Factor (APF) 10
Certifications (NIOSH) NIOSH 42 CFR 84, P95, approval number: TC-84A-1166
Certifications (OSHA) OSHA 1910.134
Color White
Design Type Cup/Molded
Disposable Single-use
Filter Type P95
Material Polypropylene, Polyester
Product Type General Purpose Particulate Respirator
Size Standard
With Exhalation Valve Yes
Filter Efficiency 95%
For Use With (Application) Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, sanding, welding
Latex Free Yes
Material Headstrap Polyisoprene, Polyester
Model 8271
Nose Piece Material Aluminum
Nosepiece Type M-nose clip
Quantity 80/Cs.
With Faceseal No
Weight (English) 0.1 lb.
Weight (Metric) 0.045 kg
Strap Attachment Type Braided and Stapled Headbands
FDA Cleared Surgical Mask No
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.