Learn More
Enterprise Technology Solutions 3M Stripping Pad, Nylon, Low Speed, 13", 5/CT, Black
SDP
Catalog No. 5000828642
Encompass_Preferred
High Productivity Pad 7300 quickly and thoroughly removes old floor finish, dirt and buildup. Use with water-based, floor finish-stripping solutions to remove old, soiled, floor finishes and sealers. Unique, open-web construction allows stripping solution to flow through pad to rinse out dirt and reduce loading for increased pad-to-floor contact and efficient stripping. Uniform distribution of mineral abrasives throughout pad - not just on the surface - delivers impressive performance and a long, useful life. Individually separated and coated fibers help produce more consistent results. Use with rotary or automatic machine speeds of 175 to 600 RPM.
Catalog No. 50-008-28642 Supplier Enterprise Technology Solutions Supplier No. MMM08271
High Productivity Pad 7300 quickly and thoroughly removes old floor finish, dirt and buildup. Use with water-based, floor finish-stripping solutions to remove old, soiled, floor finishes and sealers. Unique, open-web construction allows stripping solution to flow through pad to rinse out dirt and reduce loading for increased pad-to-floor contact and efficient stripping. Uniform distribution of mineral abrasives throughout pad - not just on the surface - delivers impressive performance and a long, useful life. Individually separated and coated fibers help produce more consistent results. Use with rotary or automatic machine speeds of 175 to 600 RPM.