Learn More
Enterprise Technology Solutions 3M Super Glue, Single Use, Pointed Tip, .07oz., 4/PK, Clear
SDP
Catalog No. 5000829879
Encompass_Preferred
Scotch Single-use Super Glue Liquid is a fast-drying, strong adhesive designed for a wide variety of porous and nonporous surfaces. It is excellent for wood, metal, rubber, vinyl, leather, ceramics, some plastics and other surfaces. Each tube contains enough adhesive for a single application. Glue comes with a precision tip for improved control.
Catalog No. 50-008-29879 Supplier Enterprise Technology Solutions Supplier No. MMMAD114
Scotch Single-use Super Glue Liquid is a fast-drying, strong adhesive designed for a wide variety of porous and nonporous surfaces. It is excellent for wood, metal, rubber, vinyl, leather, ceramics, some plastics and other surfaces. Each tube contains enough adhesive for a single application. Glue comes with a precision tip for improved control.