Description
- For regulating the flow and temperature of compressed air to 3M™ H-Series hoods and headgear
- Vortex cooling and Vortemp™ heating assemblies include waist belt and tube with quick-disconnect fitting
These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
Specifications
Specifications
| Model | W-2862 |