Learn More
3M™ Tactical 6-S Headset
Catalog No. 19033009 Shop All 3M Products
Change view
Click to view available options
Tactical 6-S headset
Catalog No.
19-033-009 Tactical 6-S headset
1 options
Catalog No. 19-033-009 Supplier 3M™ Supplier No. 7000052860
Requires
Four AAA batteries (not included).
Electronic hearing protection for shooters
Small, compact hearing protector folds into the shape of a ball for storage in a pocket or bag. Effectively perceives and localizes weak sounds and simultaneously protects against loud noise. Noise Reduction Rating (NRR): 20dB.
  • Electronically supresses sounds over 82dB (such as gunfire) to harmless levels within five milliseconds, and amplifies normal speech and range commands
  • Separate microphone, receiver, amplifier, and volume control in each cup for stereophonic reception
  • Spring-steel headband maintains attenuation better than plastic
  • Color: Gray
  • 3M Catalog# MT15H67FB-01
Order Info
Weight: 8.8 oz.
Compliance
Meets EN 352-4 standards.
TRUSTED_SUSTAINABILITY
Specifications
Model MT15H67FB-01
Noise Reduction Rating (NRR) 20 dB