Learn More
Enterprise Technology Solutions 3M Tape, Vinyl, General Purpose, 1"x36 Yards, Red
SDP
Catalog No. 5000829530
Encompass_Preferred
General-purpose Vinyl Tape 764 can be used in noncritical applications such as color-coding; bundling and safety marking; temporary marking of parts or machines; and protecting parts, equipment or products from abrasion. The noncorrosive, abrasion-resistant tape is conformable to contours, moisture-resistant and wear-resistant. It is easy to apply. Brightly colored tape features a vinyl backing, rubber adhesive and good instant adhesion. It should not be used for electrical applications.
Catalog No. 50-008-29530 Supplier Enterprise Technology Solutions Supplier No. MMM764136RED
General-purpose Vinyl Tape 764 can be used in noncritical applications such as color-coding; bundling and safety marking; temporary marking of parts or machines; and protecting parts, equipment or products from abrasion. The noncorrosive, abrasion-resistant tape is conformable to contours, moisture-resistant and wear-resistant. It is easy to apply. Brightly colored tape features a vinyl backing, rubber adhesive and good instant adhesion. It should not be used for electrical applications.