Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ 3M™ Thinsulate™-Lined Cold Weather Gloves with Goatskin Palm

Catalog No. 17000240 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-000-240 X-Large
17-000-238 Medium
17-000-239 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-000-240 Supplier Wells Lamont™ Supplier No. 894XL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3M™ Thinsulate™-Lined Cold Weather Gloves are designed to provide excellent hand protection against extreme cold temperatures, complete with a waterproof liner and goatskin palm.

The 894 lined goatskin palm glove is designed to provide excellent hand protection against extreme cold temperatures. With 3M™ Thinsulate™ insulation, the 894 glove provides additional warmth and comfort.

  • Canvasback wrist with adjustable hook closure creates comfortable, snug fit while keeping out snow, dirt, and cold air
  • Thinsulate™ liner
  • Waterproof lining

Specifications

ANSI Abrasion Rating ANSI 0
Construction Cut and Sewn
Coating Material Uncoated
Color Yellow, Black
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Material Canvas
Product Type Cold-resistant Gloves
Size X-Large
For Use With (Application) Cold Temperature
Liner Material 3M™ Thinsulate™
Series 894
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.